siRNA Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017811.3 | NP_060281.2 | 3, 4 | 944 | ||
| XM_005251496.2 | XP_005251553.1 | 2, 3 | 877 | |||
| GenBank mRNA | AJ240087.1 | - | 3, 4 | 375 | ||
| BC004862.2 | - | 3, 4 | 551 | 3838157 | ||
| BC047584.1 | - | 3, 4 | 800 | 4827644 | ||
| CR457233.1 | - | 3, 4 | 370 | |||

UBE2R2
Genomic Map