siRNA Details
| Gene Symbol: Romo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164216.1 | NP_001157688.1 | 3 | 303 | ||
| NM_001164217.2 | NP_001157689.1 | 3 | 317 | |||
| NM_025946.6 | NP_080222.1 | 2 | 401 | |||
| GenBank mRNA | AA058111.1 | - | 3 | 303 | ||
| AK003488.1 | - | 3 | 212 | |||
| AK008300.1 | - | 3 | 238 | |||
| AY028425.1 | - | 2 | 296 | |||
| BC028749.1 | - | 3 | 198 | 1196408 | ||
| CJ221444.1 | - | 3 | 265 | |||
| EL963857.1 | - | 2 | 411 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001195490, XM_006235452, FQ211245.

Romo1
Genomic Map