siRNA Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026012.2 | NP_080288.1 | 4 | 543 | ||
| GenBank mRNA | AF534394.1 | - | 3 | 355 | ||
| AK007483.1 | - | 2 | 200 | |||
| BC039808.1 | - | 4 | 513 | 1547029 | ||
| BC069856.1 | - | 4 | 543 | 6491606 | ||

Nradd
Genomic Map