siRNA Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026601.3 | NP_080877.3 | 287 | |||
| GenBank mRNA | AA403499.1 | - | 2 | 296 | ||
| BC024619.1 | - | 275 | 4923664 | |||
| BC100401.1 | - | 251 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyi| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026601.3 | NP_080877.3 | 287 | |||
| GenBank mRNA | AA403499.1 | - | 2 | 296 | ||
| BC024619.1 | - | 275 | 4923664 | |||
| BC100401.1 | - | 251 | ||||