siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000429.1 | - | 12 | 1770 | ||
| AK225424.1 | - | 13 | 1699 | |||
| AY358088.1 | - | 12 | 1797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM161A| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000429.1 | - | 12 | 1770 | ||
| AK225424.1 | - | 13 | 1699 | |||
| AY358088.1 | - | 12 | 1797 | |||