siRNA Details
| Gene Symbol: Dopey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026700.2 | NP_080976.1 | 2 | 272 | ||
| NM_027293.1 | NP_081569.1 | 2 | 272 | |||
| GenBank mRNA | AK002808.1 | - | 2 | 202 | ||
| AK147255.1 | - | 2 | 250 | |||
| AK156170.1 | - | 2 | 246 | |||
| AK164839.1 | - | 2 | 249 | |||
| BY184348.1 | - | 2 | 272 | |||
| BY195296.1 | - | 2 | 272 | |||

Dopey2
Genomic Map