siRNA Details
Gene Symbol: Cacfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029862.4 | NP_084138.1 | 6 | 1373 | ||
GenBank mRNA | AK039867.1 | - | 6 | 2180 | ||
AK052562.1 | - | 6 | 1334 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029862.4 | NP_084138.1 | 6 | 1373 | ||
GenBank mRNA | AK039867.1 | - | 6 | 2180 | ||
AK052562.1 | - | 6 | 1334 |