siRNA Details
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029862.4 | NP_084138.1 | 4, 5 | 495 | ||
| GenBank mRNA | AK039867.1 | - | 4, 5 | 472 | ||
| AK040379.1 | - | 4, 5 | 402 | |||
| AK052562.1 | - | 4, 5 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacfd1| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029862.4 | NP_084138.1 | 4, 5 | 495 | ||
| GenBank mRNA | AK039867.1 | - | 4, 5 | 472 | ||
| AK040379.1 | - | 4, 5 | 402 | |||
| AK052562.1 | - | 4, 5 | 456 | |||