siRNA Details
| Gene Symbol: Spata31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030047.2 | NP_084323.2 | 3 | 2046 | ||
| GenBank mRNA | AK133249.1 | - | 3 | 2047 | ||
| AY842249.1 | - | 3 | 1987 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata31| Gene Symbol: Spata31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030047.2 | NP_084323.2 | 3 | 2046 | ||
| GenBank mRNA | AK133249.1 | - | 3 | 2047 | ||
| AY842249.1 | - | 3 | 1987 | |||