siRNA Details
| Gene Symbol: OR14J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030946.1 | NP_112208.1 | 1 | 637 | ||
| GenBank mRNA | BC136905.1 | - | 1 | 701 | 9020896 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR14J1| Gene Symbol: OR14J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030946.1 | NP_112208.1 | 1 | 637 | ||
| GenBank mRNA | BC136905.1 | - | 1 | 701 | 9020896 | |