siRNA Details
Gene Symbol: Olfr1344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177061.3 | NP_796035.2 | 1 | 829 | ||
GenBank mRNA | BC147050.1 | - | 1 | 895 | 9056072 | |
BC147051.1 | - | 1 | 895 | 9056049 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177061.3 | NP_796035.2 | 1 | 829 | ||
GenBank mRNA | BC147050.1 | - | 1 | 895 | 9056072 | |
BC147051.1 | - | 1 | 895 | 9056049 |