siRNA Details
| Gene Symbol: Gm10408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256501.1 | NP_001243430.1 | 5 | 1077 | ||
| Gene Symbol: D830030K20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177135.4 | NP_796109.1 | 5 | 935 | ||
| GenBank mRNA | AK082998.1 | - | 5 | 1012 | ||
| AK085928.1 | - | 5 | 936 | |||

Gm10408
Genomic Map