siRNA Details
Gene Symbol: Krtap16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183296.1 | NP_899119.1 | 1 | 12 | ||
GenBank mRNA | AK017265.1 | - | 1 | 12 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183296.1 | NP_899119.1 | 1 | 12 | ||
GenBank mRNA | AK017265.1 | - | 1 | 12 |