siRNA Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145416.1 | NP_001138888.1 | 4 | 751 | ||
| NM_001321112.1 | NP_001308041.1 | 4 | 860 | |||
| NM_001321113.1 | NP_001308042.1 | 4 | 670 | |||
| NM_001321114.1 | NP_001308043.1 | 4 | 779 | |||
| NM_033408.3 | NP_212133.1 | 4 | 870 | |||
| NM_198536.2 | NP_940938.1 | 3 | 1249 | |||
| XM_005259899.2 | XP_005259956.1 | 4 | 956 | |||
| XM_017026769.1 | XP_016882258.1 | 2 | 561 | |||
| GenBank mRNA | AK127147.1 | - | 3 | 3136 | ||
| AK291979.1 | - | 4 | 730 | |||
| AY358949.1 | - | 4 | 691 | |||
| BC064948.1 | - | 3 | 990 | 6025162 | ||
| BC091472.1 | - | 4 | 678 | 30335220 | ||
| BM451378.1 | - | 4 | 756 | |||
| CB994930.1 | - | 4 | 713 | |||

TMEM205
Genomic Map