siRNA Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145416.1 | NP_001138888.1 | 2 | 218 | ||
| NM_001321112.1 | NP_001308041.1 | 2 | 327 | |||
| NM_001321113.1 | NP_001308042.1 | 2 | 137 | |||
| NM_001321114.1 | NP_001308043.1 | 2 | 246 | |||
| NM_033408.3 | NP_212133.1 | 2 | 337 | |||
| NM_198536.2 | NP_940938.1 | 1 | 716 | |||
| XM_005259899.2 | XP_005259956.1 | 2 | 423 | |||
| GenBank mRNA | AK127147.1 | - | 2 | 2499 | ||
| AK291979.1 | - | 2 | 197 | |||
| AY358949.1 | - | 2 | 158 | |||
| BC064948.1 | - | 1 | 457 | 6025162 | ||
| BC091472.1 | - | 2 | 145 | 30335220 | ||
| BE889078.1 | - | 2 | 337 | |||
| BM352837.1 | - | 2 | 96 | |||
| BM451378.1 | - | 2 | 223 | |||
| BQ639667.1 | - | 2 | 254 | |||
| CB994930.1 | - | 2 | 172 | |||

TMEM205
Genomic Map