siRNA Details
| Gene Symbol: Ugt1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201423.2 | NP_958826.1 | 1 | 597 | ||
| GenBank mRNA | AY435130.1 | - | 1 | 597 | ||
| M34007.1 | - | 1 | 591 | |||
| Gene Symbol: Ugt1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039549.1 | NP_001034638.1 | 1 | 626 | ||
| GenBank mRNA | AF461734.1 | - | 1 | 635 | ||
| AY435132.1 | - | 1 | 591 | |||

Ugt1a2
Genomic Map