siRNA Details
| Gene Symbol: Olfr1372-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137189.1 | - | 2509 | |||
| BC055827.1 | - | 2 | 1570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1372-ps1| Gene Symbol: Olfr1372-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137189.1 | - | 2509 | |||
| BC055827.1 | - | 2 | 1570 | |||