siRNA Details
Gene Symbol: Olfr1321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207631.1 | NP_997514.1 | 1 | 423 | ||
GenBank mRNA | BC119046.1 | - | 1 | 490 | 8733795 | |
BC119381.1 | - | 1 | 490 | 8734130 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
