siRNA Details
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017841.2 | NP_060311.1 | 2 | 165 | ||
GenBank mRNA | AK000494.1 | - | 2 | 165 | ||
BC002331.2 | - | 2 | 148 | 3535474 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017841.2 | NP_060311.1 | 2 | 165 | ||
GenBank mRNA | AK000494.1 | - | 2 | 165 | ||
BC002331.2 | - | 2 | 148 | 3535474 |