siRNA Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017845.3 | NP_060315.1 | 5 | 735 | ||
| XM_017008330.1 | XP_016863819.1 | 4 | 2171 | |||
| GenBank mRNA | AK000509.1 | - | 5 | 755 | ||
| BC008371.1 | - | 5 | 795 | 4051246 | ||
| BC015145.1 | - | 5 | 739 | 4052767 | ||
| BC019826.1 | - | 5 | 761 | 4995315 | ||

COMMD8
Genomic Map