siRNA Details
| Gene Symbol: Klri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012520.2 | NP_001012538.2 | 1 | 115 | ||
| GenBank mRNA | AY324874.1 | - | 1 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klri1| Gene Symbol: Klri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012520.2 | NP_001012538.2 | 1 | 115 | ||
| GenBank mRNA | AY324874.1 | - | 1 | 115 | ||