siRNA Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012621.1 | NP_001012639.1 | 12 | 1333 | ||
| XM_006236283.2 | XP_006236345.1 | 11 | 1853 | |||
| XM_006236284.2 | XP_006236346.1 | 10 | 1131 | |||
| GenBank mRNA | AY911718.1 | - | 12 | 1333 | ||
| FQ212433.1 | - | 11 | 1839 | |||

Slc13a4
Genomic Map