siRNA Details
| Gene Symbol: Ly49s5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012749.1 | NP_001012767.1 | 7 | 1050 | ||
| GenBank mRNA | AY649832.1 | - | 7 | 1050 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49s5| Gene Symbol: Ly49s5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012749.1 | NP_001012767.1 | 7 | 1050 | ||
| GenBank mRNA | AY649832.1 | - | 7 | 1050 | ||