siRNA Details
| Gene Symbol: OR9G9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013358.2 | NP_001013376.2 | 1 | 545 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR9G9| Gene Symbol: OR9G9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013358.2 | NP_001013376.2 | 1 | 545 | ||
Set Membership: |
|