siRNA Details
| Gene Symbol: Erich6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024300.1 | NP_001019471.1 | 6 | 708 | ||
| GenBank mRNA | BC083834.1 | - | 6 | 708 | 7112954 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich6| Gene Symbol: Erich6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024300.1 | NP_001019471.1 | 6 | 708 | ||
| GenBank mRNA | BC083834.1 | - | 6 | 708 | 7112954 | |