siRNA Details
Gene Symbol: LOC499643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232520.2 | XP_006232582.1 | 4 | 839 | ||
XM_008761109.1 | XP_008759331.1 | 3 | 458 | |||
GenBank mRNA | BC085354.1 | - | 4 | 729 | 7132360 |
