siRNA Details
| Gene Symbol: LOC499643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024301.2 | NP_001019472.2 | 2 | 322 | ||
| XM_006232520.2 | XP_006232582.1 | 2 | 429 | |||
| GenBank mRNA | BC085354.1 | - | 2 | 319 | 7132360 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499643| Gene Symbol: LOC499643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024301.2 | NP_001019472.2 | 2 | 322 | ||
| XM_006232520.2 | XP_006232582.1 | 2 | 429 | |||
| GenBank mRNA | BC085354.1 | - | 2 | 319 | 7132360 | |