siRNA Details
| Gene Symbol: Prpf38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024305.2 | NP_001019476.1 | 3 | 761 | ||
| XM_006233172.1 | XP_006233234.1 | 4 | 517 | |||
| XM_008761421.1 | XP_008759643.1 | 3 | 812 | |||
| GenBank mRNA | BC079264.1 | - | 3 | 710 | 7133560 | |
| FQ210223.1 | - | 3 | 741 | |||

Prpf38b
Genomic Map