siRNA Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024311.1 | NP_001019482.1 | 8, 9 | 1149 | ||
| XM_006234340.2 | XP_006234402.1 | 8, 9 | 2255 | |||
| GenBank mRNA | BC082109.1 | - | 8, 9 | 1149 | 7135070 | |

Erich2
Genomic Map