siRNA Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024311.1 | NP_001019482.1 | 6 | 903 | ||
| XM_006234340.2 | XP_006234402.1 | 6 | 2009 | |||
| GenBank mRNA | BC082109.1 | - | 6 | 903 | 7135070 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich2| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024311.1 | NP_001019482.1 | 6 | 903 | ||
| XM_006234340.2 | XP_006234402.1 | 6 | 2009 | |||
| GenBank mRNA | BC082109.1 | - | 6 | 903 | 7135070 | |