siRNA Details
Gene Symbol: Gata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024316.1 | NP_001019487.1 | 6 | 1529 | ||
XM_008762533.1 | XP_008760755.1 | 6 | 2994 | |||
GenBank mRNA | BC085855.1 | - | 6 | 1529 | 7190097 |
