siRNA Details
| Gene Symbol: Fam71f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024324.1 | NP_001019495.1 | 7 | 1488 | ||
| XM_006236223.2 | XP_006236285.1 | 9 | 1850 | |||
| GenBank mRNA | BC079297.1 | - | 7 | 1488 | 7134497 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam71f1| Gene Symbol: Fam71f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024324.1 | NP_001019495.1 | 7 | 1488 | ||
| XM_006236223.2 | XP_006236285.1 | 9 | 1850 | |||
| GenBank mRNA | BC079297.1 | - | 7 | 1488 | 7134497 | |