siRNA Details
| Gene Symbol: Gimap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024328.2 | NP_001019499.1 | 2 | 351 | ||
| GenBank mRNA | AJ633682.1 | - | 790 | |||
| BC091210.1 | - | 2 | 297 | 7381387 | ||
| CV075794.1 | - | 2 | 351 | |||
| DQ125348.1 | - | 2 | 374 | |||
| DQ125349.1 | - | 2 | 374 | |||
| FQ232730.1 | - | 2 | 383 | |||

Gimap7
Genomic Map