siRNA Details
| Gene Symbol: CLN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017882.2 | NP_060352.1 | 7 | 1155 | ||
| GenBank mRNA | AK000568.1 | - | 7 | 1100 | ||
| AK027604.1 | - | 7 | 1123 | |||
| AK074753.1 | - | 4 | 649 | |||
| AK291175.1 | - | 7 | 1082 | |||
| AK293155.1 | - | 5 | 799 | |||
| AK293197.1 | - | 7 | 1331 | |||
| BC010849.1 | - | 7 | 1069 | 3878776 | ||
| BC013130.1 | - | 7 | 1006 | 4517748 | ||

CLN6
Genomic Map