siRNA Details
| Gene Symbol: Arl8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024332.1 | NP_001019503.1 | 7 | 2550 | ||
| GenBank mRNA | BC081947.1 | - | 7 | 2550 | 7121584 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl8b| Gene Symbol: Arl8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024332.1 | NP_001019503.1 | 7 | 2550 | ||
| GenBank mRNA | BC081947.1 | - | 7 | 2550 | 7121584 | |