siRNA Details
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024335.1 | NP_001019506.1 | 6 | 904 | ||
| XM_006237380.2 | XP_006237442.1 | 5 | 2635 | |||
| XM_008763293.1 | XP_008761515.1 | 6 | 2430 | |||
| GenBank mRNA | BC095844.1 | - | 6 | 904 | 7389613 | |

Cd27
Genomic Map