siRNA Details
| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024340.1 | NP_001019511.2 | 7 | 702 | ||
| XM_006237781.2 | XP_006237843.1 | 6 | 3528 | |||
| XM_006237782.2 | XP_006237844.1 | 7 | 1331 | |||
| XM_006237784.2 | XP_006237846.1 | 7 | 3083 | |||
| XM_006237785.2 | XP_006237847.1 | 6 | 3530 | |||
| XM_006237787.2 | XP_006237849.1 | 6 | 3527 | |||
| XM_006237788.2 | XP_006237850.1 | 6 | 3525 | |||
| XM_006237789.2 | XP_006237851.1 | 6 | 3525 | |||
| GenBank mRNA | BC083882.1 | - | 7 | 702 | 7131459 | |

Mcmdc2
Genomic Map