siRNA Details
Gene Symbol: Dnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024342.1 | NP_001019513.1 | 4 | 311 | ||
GenBank mRNA | BC083662.1 | - | 4 | 311 | 7135392 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024342.1 | NP_001019513.1 | 4 | 311 | ||
GenBank mRNA | BC083662.1 | - | 4 | 311 | 7135392 |