siRNA Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024347.1 | NP_001019518.1 | 2 | 630 | ||
| XM_008764060.1 | XP_008762282.1 | 1 | 939 | |||
| XM_008764061.1 | XP_008762283.1 | 3 | 2164 | |||
| XM_008764062.1 | XP_008762284.1 | 3 | 2164 | |||
| XM_008764063.1 | XP_008762285.1 | 1 | 939 | |||
| GenBank mRNA | BC079280.1 | - | 2 | 630 | 7134187 | |

Slfnl1
Genomic Map