siRNA Details
| Gene Symbol: LOC500567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024348.1 | NP_001019519.1 | 2 | 1395 | ||
| GenBank mRNA | BC087049.1 | - | 2 | 1395 | 7112813 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500567| Gene Symbol: LOC500567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024348.1 | NP_001019519.1 | 2 | 1395 | ||
| GenBank mRNA | BC087049.1 | - | 2 | 1395 | 7112813 | |