siRNA Details
Gene Symbol: LOC500567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024348.1 | NP_001019519.1 | 2 | 1505 | ||
GenBank mRNA | BC087049.1 | - | 2 | 1505 | 7112813 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC500567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024348.1 | NP_001019519.1 | 2 | 1505 | ||
GenBank mRNA | BC087049.1 | - | 2 | 1505 | 7112813 |