siRNA Details
| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024351.1 | NP_001019522.1 | 2 | 191 | ||
| GenBank mRNA | BC088170.1 | - | 2 | 191 | 7322324 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apof| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024351.1 | NP_001019522.1 | 2 | 191 | ||
| GenBank mRNA | BC088170.1 | - | 2 | 191 | 7322324 | |