siRNA Details
| Gene Symbol: LOC500827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024352.1 | NP_001019523.2 | 4 | 622 | ||
| XM_006241290.2 | XP_006241352.1 | 4 | 488 | |||
| XM_006241291.2 | XP_006241353.1 | 5 | 1120 | |||
| XM_008765319.1 | XP_008763541.1 | 4 | 1141 | |||
| GenBank mRNA | BC078911.1 | - | 4 | 622 | 7112362 | |

LOC500827
Genomic Map