siRNA Details
| Gene Symbol: Lrrc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024354.1 | NP_001019525.1 | 4 | 1932 | ||
| XM_006241860.2 | XP_006241922.1 | 4 | 2187 | |||
| XM_006241861.1 | XP_006241923.1 | 4 | 1820 | |||
| XM_006241862.2 | XP_006241924.1 | 5 | 2705 | |||
| XM_006241865.1 | XP_006241927.1 | 5 | 1915 | |||
| GenBank mRNA | BC092583.1 | - | 4 | 1932 | 7376401 | |

Lrrc14
Genomic Map