siRNA Details
Gene Symbol: H1fnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024356.1 | NP_001019527.1 | 1 | 1118 | ||
GenBank mRNA | BC085950.1 | - | 1 | 1118 | 7132484 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: H1fnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024356.1 | NP_001019527.1 | 1 | 1118 | ||
GenBank mRNA | BC085950.1 | - | 1 | 1118 | 7132484 |