siRNA Details
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024358.1 | NP_001019529.2 | 2, 3 | 216 | ||
| GenBank mRNA | BC083665.1 | - | 2, 3 | 216 | 7135489 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glb1l3| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024358.1 | NP_001019529.2 | 2, 3 | 216 | ||
| GenBank mRNA | BC083665.1 | - | 2, 3 | 216 | 7135489 | |