siRNA Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024363.1 | NP_001019534.2 | 12, 13 | 1320 | ||
| GenBank mRNA | BC083917.1 | - | 12, 13 | 1320 | 7134752 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2cr11| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024363.1 | NP_001019534.2 | 12, 13 | 1320 | ||
| GenBank mRNA | BC083917.1 | - | 12, 13 | 1320 | 7134752 | |