siRNA Details
| Gene Symbol: Tas2r138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024685.1 | NP_001019856.1 | 1 | 958 | ||
| GenBank mRNA | AY916512.1 | - | 1 | 958 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r138| Gene Symbol: Tas2r138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024685.1 | NP_001019856.1 | 1 | 958 | ||
| GenBank mRNA | AY916512.1 | - | 1 | 958 | ||