siRNA Details
| Gene Symbol: LOC499602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025039.2 | NP_001020210.2 | 13 | 2044 | ||
| XM_006232310.2 | XP_006232372.1 | 12 | 2174 | |||
| XM_006232311.2 | XP_006232373.1 | 9 | 1711 | |||
| XM_008760940.1 | XP_008759162.1 | 12 | 1903 | |||
| GenBank mRNA | BC086604.1 | - | 3 | 1186 | 7317121 | |

LOC499602
Genomic Map