siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025046.1 | NP_001020217.2 | 7 | 1891 | ||
| XM_008762903.1 | XP_008761125.1 | 4 | 1543 | |||
| GenBank mRNA | BC092642.1 | - | 7 | 1891 | 7318030 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam131b| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025046.1 | NP_001020217.2 | 7 | 1891 | ||
| XM_008762903.1 | XP_008761125.1 | 4 | 1543 | |||
| GenBank mRNA | BC092642.1 | - | 7 | 1891 | 7318030 | |